Assay Details
Target Gene Details
Entrez Gene ID: | 20513 |
Gene Name: | solute carrier family 1 (high affinity aspartate/glutamate transporter), member 6 |
Gene Aliases: |
EAAT4 |
Location: |
Chr.10:78780496-78814825 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc1a6 | NM_009200.3 | 7 | 1279 | NP_033226.1 |
| AK159197.1 | 7 | 1280 | BAE34892.1 | |
| BC138120.1 | 7 | 1181 | ||
| BC138140.1 | 7 | 1181 | ||
| D83262.1 | 7 | 1273 | BAA23640.1 |
More Information
Set Membership: |
|

Genomic Map