Assay Details
Target Gene Details
Entrez Gene ID: | 18674 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 3 |
Gene Aliases: |
5730556H19Rik, PTP, Phc |
Location: |
Chr.10:91116578-91124052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a3 | NM_133668.3 | NP_598429.1 | ||
| AK028313.1 | ||||
| AK077274.1 | BAC36723.1 | |||
| AK077723.1 | BAC36982.1 | |||
| AK088013.1 | BAC40095.1 | |||
| AK150641.1 | BAE29729.1 | |||
| AK150729.1 | BAE29806.1 | |||
| AK150870.1 | BAE29921.1 | |||
| AK151088.1 | BAE30101.1 | |||
| AK151518.1 | BAE30467.1 | |||
| AK151887.1 | BAE30772.1 | |||
| AK152006.1 | BAE30870.1 | |||
| AK152292.1 | BAE31101.1 | |||
| AK152341.1 | BAE31137.1 | |||
| AK152713.1 | ||||
| AK167197.1 | BAE39327.1 | |||
| AK168129.1 | BAE40098.1 | |||
| AK168580.1 | BAE40449.1 | |||
| AK203521.1 | ||||
| BC018161.1 | AAH18161.1 |
More Information
Set Membership: |
|

Genomic Map