Assay Details
Target Gene Details
Entrez Gene ID: | 18674 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 3 |
Gene Aliases: |
5730556H19Rik, PTP, Phc |
Location: |
Chr.10:91116578-91124052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
More Information
Set Membership: |
|

Genomic Map