Assay Details
Target Gene Details
Entrez Gene ID: | 237553 |
Gene Name: | TRH-degrading enzyme |
Gene Aliases: |
9330155P21Rik |
Location: |
Chr.10:114398821-114801370 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trhde | NM_146241.2 | 4 | 1332 | NP_666353.1 |
| AK034093.1 | 4 | 711 | BAC28583.1 | |
| BC032288.1 | 4 | 1337 | AAH32288.1 |
More Information
Set Membership: |
|

Genomic Map