Assay Details
Target Gene Details
Entrez Gene ID: | 67418 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 4 |
Gene Aliases: |
3732410E19Rik, 3830425H19Rik, AI788954, AW146233, PPIase |
Location: |
Chr.10:7792891-7822563 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppil4 | NM_026141.3 | NP_080417.2 | ||
| AK011443.1 | BAB27623.1 | |||
| AK012371.1 | BAB28194.1 | |||
| AK014453.1 | ||||
| AK017370.1 | BAB30711.1 | |||
| AK049634.1 | ||||
| AK164127.1 | ||||
| AK167168.1 | ||||
| AK167258.1 | BAE39374.1 | |||
| BC079912.1 | AAH79912.1 |
More Information
Set Membership: |
|

Genomic Map