Assay Details
Target Gene Details
Entrez Gene ID: | 20224 |
Gene Name: | SAR1 gene homolog A (S. cerevisiae) |
Gene Aliases: |
1600019H17Rik, Sara, Sara1 |
Location: |
Chr.10:61680304-61693297 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sar1a | NM_009120.2 | NP_033146.1 | ||
| AK157843.1 | BAE34224.1 | |||
| AK159239.1 | BAE34922.1 | |||
| AK168175.1 | BAE40135.1 | |||
| BC005549.1 | AAH05549.1 | |||
| L20294.1 | AAA16323.1 |

Genomic Map