Assay Details
Target Gene Details
Entrez Gene ID: | 56480 |
Gene Name: | TANK-binding kinase 1 |
Gene Aliases: |
1200008B05Rik, AI462036, AW048562 |
Location: |
Chr.10:121546455-121586794 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbk1 | NM_019786.4 | 1 | 54 | NP_062760.3 |
| AK004649.2 | BAB23440.1 | |||
| AK083214.1 | ||||
| AK168193.1 | 1 | 40 | BAE40153.1 | |
| BU708441.1 | 1 | 47 |
More Information
Set Membership: |
|

Genomic Map