Assay Details
Target Gene Details
Entrez Gene ID: | 56351 |
Gene Name: | prostaglandin E synthase 3 (cytosolic) |
Gene Aliases: |
5730442A20Rik, Ptges, Tebp, cPGES, p23, sid3177 |
Location: |
Chr.10:128058982-128077254 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptges3 | NM_019766.4 | NP_062740.1 | ||
| AB024935.1 | BAA84684.1 | |||
| AF153479.1 | AAD39543.1 | |||
| AK008805.1 | BAB25906.1 | |||
| AK010636.1 | BAB27080.2 | |||
| AK017742.1 | ||||
| AK075932.1 | BAC36062.1 | |||
| AK075987.1 | BAC36099.1 | |||
| AK077538.1 | BAC36854.1 | |||
| AK167443.1 | BAE39529.1 | |||
| AK168073.1 | BAE40047.1 | |||
| AK168210.1 | BAE40169.1 | |||
| AK168721.1 | BAE40563.1 | |||
| AY281130.1 | AAP34198.1 | |||
| BC003708.1 | AAH03708.1 | |||
| BC085264.1 | AAH85264.1 |
More Information
Set Membership: |
|

Genomic Map