Assay Details
Target Gene Details
Entrez Gene ID: | 56533 |
Gene Name: | regulator of G-protein signaling 17 |
Gene Aliases: |
6430507P11Rik, Rgsz2 |
Location: |
Chr.10:5825663-5922400 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs17 | NM_001161822.1 | 1 | 39 | NP_001155294.1 |
| NM_019958.4 | 1 | 39 | NP_064342.1 | |
| AF191555.1 | AAF05758.1 | |||
| AK018279.1 | BAB31145.1 | |||
| AK030492.1 | 1 | 40 | BAC26989.1 | |
| AK045770.1 | BAC32486.1 | |||
| AK046009.1 | BAC32571.1 | |||
| AK078745.1 | BAC37375.1 | |||
| AK163188.1 | BAE37226.1 | |||
| BC064782.1 | AAH64782.1 |
More Information
Set Membership: |
|

Genomic Map