Assay Details
Target Gene Details
Entrez Gene ID: | 338359 |
Gene Name: | suppressor of var1, 3-like 1 (S. cerevisiae) |
Gene Aliases: |
6330443E10Rik |
Location: |
Chr.10:62429329-62451294 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supv3l1 | NM_181423.2 | 5 | 630 | NP_852088.1 |
| AJ968954.1 | 5 | 657 | CAI92124.1 | |
| AK031911.1 | 5 | 621 | ||
| AK143755.1 | 5 | 655 | ||
| BC049796.1 | 5 | 630 | AAH49796.1 |
More Information
Set Membership: |
|

Genomic Map