Assay Details
Target Gene Details
Entrez Gene ID: | 29863 |
Gene Name: | phosphodiesterase 7B |
Gene Aliases: |
- |
Location: |
Chr.10:20397052-20725068 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde7b | NM_013875.5 | 10 | 1248 | NP_038903.3 |
| AF190639.1 | 10 | 1029 | AAF25195.1 | |
| AJ251859.1 | 10 | 1300 | CAB92530.1 | |
| AK035385.1 | 10 | 1260 | BAC29052.1 | |
| AK049910.1 | 10 | 1304 | ||
| BC130267.1 | 10 | 1129 |
More Information
Set Membership: |
|

Genomic Map