Assay Details
Target Gene Details
Entrez Gene ID: | 237422 |
Gene Name: | resistance to inhibitors of cholinesterase 8 homolog B (C. elegans) |
Gene Aliases: |
BC051080, Ric-8, Ric-8b |
Location: |
Chr.10:84917249-85018337 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ric8b | NM_001013441.2 | NP_001013459.1 | ||
| NM_183172.2 | NP_898995.1 | |||
| AK132369.1 | BAE21130.1 | |||
| AK137531.1 | ||||
| AY940666.1 | AAX46315.1 | |||
| AY940667.1 | AAX46316.1 | |||
| BC051080.1 | AAH51080.1 |

Genomic Map