Assay Details
Target Gene Details
Entrez Gene ID: | 338365 |
Gene Name: | solute carrier family 41, member 2 |
Gene Aliases: |
A230035L05Rik, SLC41A1-L1 |
Location: |
Chr.10:83230848-83338329 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc41a2 | NM_177388.3 | NP_796362.1 | ||
| AK038549.1 | BAC30038.1 | |||
| BC138146.1 | ||||
| BC138147.1 |
More Information
Set Membership: |
|

Genomic Map