Assay Details
Target Gene Details
Entrez Gene ID: | 70423 |
Gene Name: | tetraspanin 15 |
Gene Aliases: |
1110036D12Rik, 2700063A19Rik, AW048364, Tm4sf15 |
Location: |
Chr.10:62185396-62231277 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan15 | NM_197996.2 | NP_932113.2 | ||
| AK012471.1 | ||||
| AK172169.1 | BAE42863.1 | |||
| CN455615.1 | ||||
| CX206586.1 |
More Information
Set Membership: |
|

Genomic Map