Assay Details
Target Gene Details
Entrez Gene ID: | 19072 |
Gene Name: | prolyl endopeptidase |
Gene Aliases: |
AI047692, AI450383, D10Wsu136e, PEP, Pop |
Location: |
Chr.10:45067214-45159981 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prep | NM_011156.2 | 10 | 1384 | NP_035286.1 |
| AB007631.1 | 10 | 1267 | BAA88239.1 | |
| AK053816.1 | 10 | 1395 | BAC35538.1 | |
| AK076257.1 | 10 | 1383 | BAC36278.1 | |
| AK133875.1 | 10 | 1383 | ||
| AK141851.1 | 10 | 1393 | BAE24858.1 | |
| AK161528.1 | 10 | 1383 | BAE36445.1 | |
| AK163774.1 | 2 | 260 | BAE37487.1 | |
| AK163787.1 | 10 | 1396 | BAE37495.1 | |
| AK170570.1 | 10 | 1393 | BAE41885.1 | |
| AK210695.1 | 2 | 200 | ||
| BC012869.1 | 10 | 1384 | AAH12869.1 | |
| BC050830.1 | 10 | 1377 | AAH50830.2 |
More Information
Set Membership: |
|

Genomic Map