Assay Details
Target Gene Details
Entrez Gene ID: | 237523 |
Gene Name: | protein tyrosine phosphatase, receptor type, Q |
Gene Aliases: |
- |
Location: |
Chr.10:107517360-107720078 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprq | NM_001081432.1 | NP_001074901.1 |
More Information
Set Membership: |
|

Genomic Map