Assay Details
Target Gene Details
Entrez Gene ID: | 64452 |
Gene Name: | solute carrier family 5, member 4a |
Gene Aliases: |
SGLT3-a |
Location: |
Chr.10:76147451-76189265 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a4a | NM_133184.2 | NP_573447.2 | ||
| AF251267.1 | AAG01741.1 |
More Information
Set Membership: |
|

Genomic Map