Assay Details
Target Gene Details
Entrez Gene ID: | 432478 |
Gene Name: | transmembrane protease, serine 9 |
Gene Aliases: |
Serase-1B |
Location: |
Chr.10:80871117-80899495 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss9 | NM_001081688.2 | 1 | 77 | NP_001075157.2 |
| AB109391.1 | 1 | 188 | ||
| BC158102.1 | 1 | 173 | ||
| BC172169.1 | 1 | 173 |
More Information
Set Membership: |
|

Genomic Map