Assay Details
Target Gene Details
Entrez Gene ID: | 56516 |
Gene Name: | RNA binding motif, single stranded interacting protein 2 |
Gene Aliases: |
2610315E04Rik, Scr3 |
Location: |
Chr.10:128129469-128180297 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbms2 | NM_001039080.1 | NP_001034169.1 | ||
| NM_019711.2 | NP_062685.2 | |||
| AB026583.1 | BAA77263.1 | |||
| AK054482.1 | BAC35797.1 | |||
| AK147838.1 | BAE28173.1 | |||
| BC021627.1 | AAH21627.1 | |||
| CX564775.1 |

Genomic Map