Assay Details
Target Gene Details
Entrez Gene ID: | 64454 |
Gene Name: | solute carrier family 5 (neutral amino acid transporters, system A), member 4b |
Gene Aliases: |
2010104G07Rik, SAAT1, SGLT3-b, SGLT3b, pSGLT2 |
Location: |
Chr.10:76058618-76111018 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a4b | NM_023219.2 | 5 | 513 | NP_075708.2 |
| AF251268.1 | 5 | 513 | AAG01742.1 | |
| AF411960.2 | 5 | 433 | AAL06342.2 | |
| BC109133.1 | 5 | 452 | AAI09134.1 | |
| BC109134.1 | 5 | 452 | AAI09135.1 | |
| BC127612.1 | 5 | 452 | ||
| BC127615.1 | 5 | 452 | ||
| BC128465.1 | 5 | 452 |
More Information
Set Membership: |
|

Genomic Map