Assay Details
Target Gene Details
Entrez Gene ID: | 50493 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
TR, TR1, TrxR1 |
Location: |
Chr.10:82833951-82897724 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Txnrd1 | NM_001042523.1 | NP_001035988.1 | ||
| AF333036.1 | AAK01140.1 |
More Information
Set Membership: |
|

Genomic Map