Assay Details
Target Gene Details
Entrez Gene ID: | 19279 |
Gene Name: | protein tyrosine phosphatase, receptor type, R |
Gene Aliases: |
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213 |
Location: |
Chr.10:116018200-116274932 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprr | NM_001161837.1 | 2 | 196 | NP_001155309.1 |
NM_011217.2 | 3 | 802 | NP_035347.1 | |
AK028054.1 | 3 | 803 | BAC25724.1 | |
BC119230.1 | 3 | 735 | ||
BC120547.1 | 3 | 735 | ||
CJ061226.1 | 2 | 196 | ||
D31898.1 | 3 | 765 | BAA06696.1 | |
Z30313.1 | 2 | 191 | CAA82957.1 |
