Assay Details
Target Gene Details
Entrez Gene ID: | 18114 |
Gene Name: | ribosomal RNA processing 1 homolog (S. cerevisiae) |
Gene Aliases: |
AL033305, AL033306, AU018604, NNP-1, Nnp1 |
Location: |
Chr.10:78400362-78413043 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrp1 | NM_010925.2 | 1 | 68 | NP_035055.2 |
| AK077540.1 | ||||
| BC022910.1 | ||||
| BC065993.1 | AAH65993.1 | |||
| BC065994.1 | AAH65994.1 | |||
| BY106122.1 | 1 | 68 | ||
| U79773.1 | AAC53286.1 | |||
| U79774.1 | AAB63261.1 |

Genomic Map