Assay Details
Target Gene Details
Entrez Gene ID: | 327744 |
Gene Name: | RIKEN cDNA E130307A14 gene |
Gene Aliases: |
- |
Location: |
Chr.10:39621411-39732007 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E130307A14Rik | NR_038037.1 | |||
| AK164126.1 |
Target Gene Details
Entrez Gene ID: | 103213 |
Gene Name: | TRAF3 interacting protein 2 |
Gene Aliases: |
AI429613, Act1, CIKS |
Location: |
Chr.10:39612934-39655307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf3ip2 | NM_134000.3 | NP_598761.2 | ||
| AB238206.1 | ||||
| AB238207.1 | ||||
| AK031556.1 | BAC27449.1 | |||
| AK037407.1 | BAC29801.1 | |||
| AK052190.1 | BAC34875.1 | |||
| AK098107.1 | BAC05236.1 | |||
| BC022633.1 | AAH22633.1 | |||
| BC071207.1 | AAH71207.1 | |||
| BC096483.1 | AAH96483.1 | |||
| BC138234.1 | ||||
| BC138235.1 | ||||
| JX869941.1 |
More Information
Set Membership: |
|

Genomic Map