Assay Details
Target Gene Details
Entrez Gene ID: | 67895 |
Gene Name: | pyrophosphatase (inorganic) 1 |
Gene Aliases: |
2010317E03Rik, Pyp |
Location: |
Chr.10:61648621-61674165 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppa1 | NM_026438.4 | NP_080714.2 | ||
| AK008575.1 | BAB25754.1 | |||
| AK020291.1 | ||||
| AK088397.1 | BAC40327.1 | |||
| AK145640.1 | BAE26558.1 | |||
| AK146300.1 | BAE27057.1 | |||
| AK151512.1 | BAE30462.1 | |||
| BC010468.1 | AAH10468.1 | |||
| CT010203.1 | CAJ18411.1 |
More Information
Set Membership: |
|

Genomic Map