Assay Details
Target Gene Details
Entrez Gene ID: | 75089 |
Gene Name: | UHRF1 (ICBP90) binding protein 1-like |
Gene Aliases: |
2010319N22Rik, 4930506D01Rik, AA589495, C80672, E030041M21Rik, mKIAA0701 |
Location: |
Chr.10:89744991-89819891 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf1bp1l | NM_029166.2 | NP_083442.2 | ||
AB093261.1 | BAC41445.1 | |||
AK033300.1 | ||||
AK087280.1 | BAC39836.1 | |||
AK141050.1 | ||||
BB529788.2 | ||||
BC132134.1 | ||||
BC138092.1 |
