Assay Details
Target Gene Details
Entrez Gene ID: | 20393 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
Sgk |
Location: |
Chr.10:21882184-21999903 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgk1 | NM_001161845.2 | 6 | 1015 | NP_001155317.2 |
| NM_001161847.2 | 4 | 516 | NP_001155319.1 | |
| NM_001161848.2 | 4 | 309 | NP_001155320.1 | |
| NM_001161849.2 | 4 | 390 | NP_001155321.1 | |
| NM_001161850.2 | 4 | 497 | NP_001155322.1 | |
| NM_011361.3 | 4 | 365 | NP_035491.1 | |
| AF139638.1 | 4 | 349 | AAD43302.1 | |
| AF205855.1 | 4 | 346 | AAF19429.1 | |
| AK086892.1 | 4 | 349 | ||
| AK132234.1 | 4 | 437 | BAE21048.1 | |
| AK146037.1 | 4 | 349 | BAE26849.1 | |
| AK146062.1 | 4 | 346 | BAE26871.1 | |
| AK159131.1 | 4 | 349 | BAE34843.1 | |
| AK167364.1 | 4 | 346 | BAE39461.1 | |
| BC005720.1 | 4 | 326 | AAH05720.1 | |
| BC070401.1 | 6 | 1015 | AAH70401.1 | |
| BY736002.1 | 4 | 309 | ||
| CK626189.1 | 4 | 497 | ||
| CN709375.1 | 4 | 233 |
More Information
Set Membership: |
|

Genomic Map