Assay Details
Target Gene Details
Entrez Gene ID: | 66456 |
Gene Name: | RIKEN cDNA 2810001G20 gene |
Gene Aliases: |
AW045514 |
Location: |
Chr.11:64079484-64083259 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2810001G20Rik | NR_033780.1 | |||
| AK005116.1 | ||||
| AK007927.1 | ||||
| AK012631.1 |
Target Gene Details
Entrez Gene ID: | 70383 |
Gene Name: | cytochrome c oxidase assembly protein 10 |
Gene Aliases: |
2410004F01Rik, AU042636 |
Location: |
Chr.11:63962627-64079472 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox10 | NM_178379.3 | NP_848466.1 | ||
| AK154668.1 | BAE32754.1 | |||
| AK156865.1 | BAE33878.1 | |||
| AK172718.1 | BAE43143.1 | |||
| BY078593.1 |
More Information
Set Membership: |
|

Genomic Map