Assay Details
Target Gene Details
Entrez Gene ID: | 66456 |
Gene Name: | RIKEN cDNA 2810001G20 gene |
Gene Aliases: |
AW045514 |
Location: |
Chr.11:64079484-64083259 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2810001G20Rik | NR_033780.1 | 1 | 251 | |
| AK005116.1 | 1 | 218 | ||
| AK007927.1 | 1 | 253 | ||
| AK012631.1 | 1 | 244 |

Genomic Map