Assay Details
Target Gene Details
Entrez Gene ID: | 217262 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 9 |
Gene Aliases: |
D630040K07Rik |
Location: |
Chr.11:110100822-110168153 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca9 | NM_147220.2 | NP_671753.2 | ||
| AF491299.1 | AAM90894.1 | |||
| AK029256.1 | BAC26358.1 |
More Information
Set Membership: |
|

Genomic Map