Assay Details
Target Gene Details
Entrez Gene ID: | 237911 |
Gene Name: | BRCA1 interacting protein C-terminal helicase 1 |
Gene Aliases: |
3110009N10Rik, 8030460J03Rik, Bach1, FACJ, Fancj, OF |
Location: |
Chr.11:86058131-86201267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brip1 | NM_178309.2 | NP_840094.1 | ||
| AK033205.1 | ||||
| AK041841.1 | BAE20606.1 | |||
| AK080771.1 | BAC38016.1 | |||
| AK145709.1 | BAE26603.1 | |||
| AK169440.1 | BAE41179.1 | |||
| AK169823.1 | BAE41393.1 | |||
| BC094252.1 | AAH94252.1 |
More Information
Set Membership: |
|

Genomic Map