Assay Details
Target Gene Details
Entrez Gene ID: | 73324 |
Gene Name: | clathrin heavy chain linker domain containing 1 |
Gene Aliases: |
1700034F02Rik |
Location: |
Chr.11:29547950-29578360 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clhc1 | NM_001163521.1 | NP_001156993.1 | ||
| NM_028504.1 | NP_082780.1 | |||
| AK006591.1 | BAB24663.1 | |||
| BC137683.1 | ||||
| BC144828.1 |
More Information
Set Membership: |
|

Genomic Map