Assay Details
Target Gene Details
Entrez Gene ID: | 11302 |
Gene Name: | apoptosis-associated tyrosine kinase |
Gene Aliases: |
AATYK, aatyk1, mKIAA0641 |
Location: |
Chr.11:120007311-120047182 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aatk | NM_001198785.1 | NP_001185714.1 | ||
| NM_007377.4 | 1 | 414 | NP_031403.2 | |
| AB093253.2 | 1 | 414 | BAC41437.2 | |
| AB288871.1 | ||||
| AY236858.1 | AAO92350.1 | |||
| AY236859.1 | AAO92351.1 | |||
| BC080846.1 | AAH80846.1 |
More Information
Set Membership: |
|

Genomic Map