Assay Details
Target Gene Details
Entrez Gene ID: | 11488 |
Gene Name: | a disintegrin and metallopeptidase domain 11 |
Gene Aliases: |
AW060611, Mdc |
Location: |
Chr.11:102761439-102780262 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam11 | NM_001110778.1 | 23 | 2138 | NP_001104248.1 |
| NM_009613.2 | 23 | 2138 | NP_033743.2 | |
| AB009676.1 | 23 | 1971 | BAA83384.1 | |
| AK157926.1 | 23 | 2143 | BAE34267.1 | |
| BC054536.1 | 23 | 1998 | AAH54536.1 |
More Information
Set Membership: |
|

Genomic Map