Assay Details
Target Gene Details
Entrez Gene ID: | 108100 |
Gene Name: | brain-specific angiogenesis inhibitor 1-associated protein 2 |
Gene Aliases: |
IRSp53, R75030 |
Location: |
Chr.11:119942869-120006782 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baiap2 | NM_001037754.3 | 14 | 1658 | NP_001032843.1 |
| NM_001037755.3 | NP_001032844.2 | |||
| NM_130862.4 | 14 | 1778 | NP_570932.2 | |
| AF390178.1 | AAK68153.1 | |||
| AF390179.1 | AAK81838.1 | |||
| AK049469.1 | BAC33764.1 | |||
| AK145924.1 | 14 | 1779 | BAE26755.1 | |
| AK158579.1 | 14 | 1800 | ||
| AK160401.1 | 14 | 1660 | BAE35770.1 | |
| AK170190.1 | 7 | 1307 | ||
| AK186120.1 | ||||
| BC006620.1 | 13 | 2256 | AAH06620.1 | |
| BC016411.1 | 13 | 2256 | AAH16411.1 |
More Information
Set Membership: |
|

Genomic Map