Assay Details
Target Gene Details
Entrez Gene ID: | 73635 |
Gene Name: | prostaglandin E synthase 3 (cytosolic)-like |
Gene Aliases: |
1110069E20Rik, 1700113I22Rik |
Location: |
Chr.11:101418801-101425333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptges3l | NM_026865.2 | NP_081141.1 | ||
| AK004415.1 | BAB23297.1 | |||
| AK007198.1 | BAB24896.1 | |||
| BC141198.1 |
More Information
Set Membership: |
|

Genomic Map