Assay Details
Target Gene Details
Entrez Gene ID: | 73634 |
Gene Name: | RIKEN cDNA 1700125H20 gene |
Gene Aliases: |
- |
Location: |
Chr.11:85140088-85181164 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700125H20Rik | NM_028589.1 | 3 | 283 | NP_082865.1 |
| AK007281.1 | BAB24934.1 | |||
| BC048674.1 | 3 | 283 | AAH48674.1 |

Genomic Map