Assay Details
Target Gene Details
Entrez Gene ID: | 11492 |
Gene Name: | a disintegrin and metallopeptidase domain 19 (meltrin beta) |
Gene Aliases: |
AL024287, Mltnb |
Location: |
Chr.11:46054502-46147347 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam19 | NM_001291890.1 | NP_001278819.1 | ||
| NM_001291891.1 | NP_001278820.1 | |||
| NM_009616.4 | NP_033746.1 | |||
| AF019887.2 | AAC40037.1 | |||
| AK147217.1 | BAE27774.1 | |||
| AK147549.1 | BAE27990.1 | |||
| BC115767.1 | ||||
| CJ111383.1 | ||||
| CJ111483.1 | ||||
| CJ114671.1 | ||||
| D50410.2 | BAA18923.2 |
More Information
Set Membership: |
|

Genomic Map