Assay Details
Target Gene Details
Entrez Gene ID: | 103135 |
Gene Name: | PAN2 polyA specific ribonuclease subunit homolog (S. cerevisiae) |
Gene Aliases: |
1200014O24Rik, AI047843, AW742773, Usp52, mKIAA0710 |
Location: |
Chr.10:128303331-128321358 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pan2 | NM_001252326.1 | 23 | 3405 | NP_001239255.1 |
| NM_001252327.1 | 23 | 3351 | NP_001239256.1 | |
| NM_133992.3 | 23 | 3432 | NP_598753.1 | |
| AK004773.2 | 22 | 3431 | ||
| AK047887.1 | 23 | 3392 | BAC33183.1 | |
| AK089001.1 | 23 | 3433 | BAC40693.1 | |
| AK129196.1 | 21 | 3902 | BAC98006.2 | |
| AK157800.1 | 23 | 3309 | BAE34203.1 | |
| AK160655.1 | 5 | 554 | ||
| BC006064.1 | 3 | 388 | AAH06064.1 | |
| BC043659.1 | 10 | 1344 | AAH43659.1 | |
| BC075686.1 | 23 | 3409 | AAH75686.1 | |
| BC079841.1 | 23 | 3366 | AAH79841.1 |
More Information
Set Membership: |
|

Genomic Map