Assay Details
Target Gene Details
Entrez Gene ID: | 103135 |
Gene Name: | PAN2 polyA specific ribonuclease subunit homolog (S. cerevisiae) |
Gene Aliases: |
1200014O24Rik, AI047843, AW742773, Usp52, mKIAA0710 |
Location: |
Chr.10:128303331-128321358 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pan2 | NM_001252326.1 | NP_001239255.1 | ||
| NM_001252327.1 | NP_001239256.1 | |||
| NM_133992.3 | NP_598753.1 | |||
| AK004773.2 | ||||
| AK047887.1 | BAC33183.1 | |||
| AK089001.1 | BAC40693.1 | |||
| AK129196.1 | BAC98006.2 | |||
| AK157800.1 | BAE34203.1 | |||
| BC075686.1 | AAH75686.1 | |||
| BC079841.1 | AAH79841.1 | |||
| BG086688.2 |
More Information
Set Membership: |
|

Genomic Map