Assay Details
Target Gene Details
Entrez Gene ID: | 65257 |
Gene Name: | ankyrin repeat and SOCS box-containing 3 |
Gene Aliases: |
2400011J03Rik, Asb-3 |
Location: |
Chr.11:30954194-31102704 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb3 | NM_023906.3 | 2 | 233 | NP_076395.2 |
| AF155354.1 | 1 | 48 | AAD38810.1 | |
| AK010315.1 | 2 | 169 | BAB26847.1 | |
| AK028489.1 | 2 | 226 | ||
| AK038086.1 | 2 | 214 | ||
| AK038897.1 | 2 | 184 | ||
| AK143580.1 | 2 | 168 | BAE25448.1 | |
| BB618690.1 | 2 | 235 | ||
| BC023086.1 | 2 | 167 | AAH23086.1 |

Genomic Map