Assay Details
Target Gene Details
Entrez Gene ID: | 66431 |
Gene Name: | oxidoreductase like domain containing 1 |
Gene Aliases: |
1810049H13Rik |
Location: |
Chr.11:120456601-120458091 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxld1 | NM_025560.2 | 1 | 168 | NP_079836.1 |
| AK007842.1 | 1 | 169 | BAB25299.1 | |
| AK021150.1 | 1 | 156 | BAB32307.1 | |
| BC147291.1 | 1 | 161 | ||
| BC147292.1 | 1 | 161 | ||
| BC172027.1 |
More Information
Set Membership: |
|

Genomic Map