Assay Details
Target Gene Details
Entrez Gene ID: | 11430 |
Gene Name: | acyl-Coenzyme A oxidase 1, palmitoyl |
Gene Aliases: |
AOX, Acox, D130055E20Rik, Paox |
Location: |
Chr.11:116171883-116199045 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acox1 | NM_001271898.1 | 5 | 940 | NP_001258827.1 |
| NM_015729.3 | 5 | 940 | NP_056544.2 | |
| AB034914.1 | 5 | 629 | BAA86870.1 | |
| AF006688.1 | 5 | 679 | AAB62926.1 | |
| AK034760.1 | 2 | 415 | ||
| AK040566.1 | 5 | 643 | BAC30628.1 | |
| AK054446.1 | 5 | 716 | BAC35782.2 | |
| AK128941.1 | 5 | 642 | ||
| AK170217.1 | BAE41642.1 | |||
| BC056448.1 | 5 | 940 | AAH56448.1 |
More Information
Set Membership: |
|

Genomic Map