Assay Details
Target Gene Details
Entrez Gene ID: | 217370 |
Gene Name: | cDNA sequence BC017643 |
Gene Aliases: |
- |
Location: |
Chr.11:121222588-121229558 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC017643 | NM_001252548.1 | NP_001239477.1 | ||
| NM_001252549.1 | NP_001239478.1 | |||
| NM_001252550.1 | NP_001239479.1 | |||
| NM_001254735.1 | NP_001241664.1 | |||
| NM_144832.2 | NP_659081.1 | |||
| AK143743.1 | 3 | 229 | ||
| AK145884.1 | BAE26724.1 | |||
| AK158402.1 | BAE34490.1 | |||
| AK159453.1 | BAE35096.1 | |||
| AK168104.1 | BAE40075.1 | |||
| AK170767.1 | BAE42017.1 | |||
| BC017643.1 | AAH17643.1 | |||
| BC026616.1 | AAH26616.1 | |||
| BC027406.1 | AAH27406.1 | |||
| BU059230.1 | ||||
| BY244471.1 | ||||
| CD806773.1 | ||||
| CN716786.1 |
More Information
Set Membership: |
|

Genomic Map