Assay Details
Target Gene Details
Entrez Gene ID: | 217370 |
Gene Name: | cDNA sequence BC017643 |
Gene Aliases: |
- |
Location: |
Chr.11:121222588-121229558 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC017643 | NM_001252548.1 | 6 | 677 | NP_001239477.1 |
| NM_001252549.1 | 6 | 523 | NP_001239478.1 | |
| NM_001252550.1 | 5 | 623 | NP_001239479.1 | |
| NM_001254735.1 | 6 | 600 | NP_001241664.1 | |
| NM_144832.2 | 6 | 697 | NP_659081.1 | |
| AK143743.1 | 4 | 1714 | ||
| AK145884.1 | 6 | 663 | BAE26724.1 | |
| AK158402.1 | 6 | 634 | BAE34490.1 | |
| AK159453.1 | 6 | 655 | BAE35096.1 | |
| AK168104.1 | 6 | 655 | BAE40075.1 | |
| AK170767.1 | 6 | 645 | BAE42017.1 | |
| AK185885.1 | ||||
| BC017643.1 | 6 | 653 | AAH17643.1 | |
| BC026616.1 | 6 | 588 | AAH26616.1 | |
| BC027406.1 | 6 | 482 | AAH27406.1 | |
| BU059230.1 | ||||
| CD806773.1 | 6 | 600 | ||
| CN716786.1 | 5 | 549 |
More Information
Set Membership: |
|

Genomic Map