Assay Details
Target Gene Details
Entrez Gene ID: | 216874 |
Gene Name: | calmodulin binding transcription activator 2 |
Gene Aliases: |
Kiaa0909-hp, mKIAA0909 |
Location: |
Chr.11:70669463-70688108 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camta2 | NM_001190376.1 | 21 | 3659 | NP_001177305.1 |
| NM_001190378.1 | 20 | 3638 | NP_001177307.1 | |
| NM_001190379.1 | 19 | 3566 | NP_001177308.1 | |
| NM_178116.4 | 21 | 3674 | NP_835217.1 | |
| AK122400.1 | 19 | 3315 | BAC65682.1 | |
| AK150186.1 | 20 | 3492 | ||
| AK169118.1 | 19 | 3405 | BAE40898.1 | |
| BC027039.1 | 10 | 1383 | AAH27039.1 | |
| BC027385.1 | AAH27385.1 | |||
| BC049133.1 | 21 | 3503 | AAH49133.1 | |
| BC056395.1 | 21 | 3488 | AAH56395.1 |
More Information
Set Membership: |
|

Genomic Map