Assay Details
Target Gene Details
Entrez Gene ID: | 11688 |
Gene Name: | arachidonate 8-lipoxygenase |
Gene Aliases: |
15-LOX-2, 15-LOX-B, 8-LOX, 8S-LOX, Alox15b |
Location: |
Chr.11:69183885-69197843 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alox8 | NM_009661.4 | NP_033791.1 | ||
| AK028724.1 | BAC26085.1 | |||
| AK158553.1 | BAE34556.1 | |||
| BC015253.1 | AAH15253.1 | |||
| U93277.1 | AAC53356.1 | |||
| Y14696.1 | CAA75003.1 |
More Information
Set Membership: |
|

Genomic Map