Assay Details
Target Gene Details
Entrez Gene ID: | 68938 |
Gene Name: | alveolar soft part sarcoma chromosome region, candidate 1 (human) |
Gene Aliases: |
1190006K01Rik, ASPC, ASPCR1, ASPL, ASPS, RCC17, TUG |
Location: |
Chr.11:120672439-120709447 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aspscr1 | NM_001164224.1 | 7 | 887 | NP_001157696.1 |
| NM_026877.2 | 8 | 1108 | NP_081153.1 | |
| NM_198223.2 | 7 | 887 | NP_937866.1 | |
| AK004509.1 | 2 | 120 | BAB23341.1 | |
| AK088197.1 | 8 | 947 | ||
| AK146061.1 | 7 | 888 | BAE26870.1 | |
| AK183906.1 | ||||
| AY349132.1 | 6 | 877 | AAR01613.1 | |
| AY349133.1 | 8 | 1025 | AAR01614.1 | |
| BC019177.1 | 8 | 1051 | AAH19177.1 | |
| BC022115.1 | 8 | 1055 | AAH22115.1 | |
| BC031153.1 | 8 | 1040 | AAH31153.1 |

Genomic Map