Assay Details
Target Gene Details
Entrez Gene ID: | 68938 |
Gene Name: | alveolar soft part sarcoma chromosome region, candidate 1 (human) |
Gene Aliases: |
1190006K01Rik, ASPC, ASPCR1, ASPL, ASPS, RCC17, TUG |
Location: |
Chr.11:120672439-120709447 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aspscr1 | NM_001164224.1 | NP_001157696.1 | ||
| NM_026877.2 | NP_081153.1 | |||
| NM_198223.2 | NP_937866.1 | |||
| AK004509.1 | BAB23341.1 | |||
| AK088197.1 | ||||
| AK146061.1 | BAE26870.1 | |||
| AY349132.1 | AAR01613.1 | |||
| AY349133.1 | AAR01614.1 | |||
| BC019177.1 | AAH19177.1 | |||
| BC022115.1 | AAH22115.1 | |||
| BC031153.1 | AAH31153.1 |

Genomic Map