Assay Details
Target Gene Details
Entrez Gene ID: | 12029 |
Gene Name: | B cell CLL/lymphoma 6, member B |
Gene Aliases: |
Bazf |
Location: |
Chr.11:70224127-70229798 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl6b | NM_007528.3 | NP_031554.1 | ||
| AK156733.1 | BAE33827.1 | |||
| BC090998.1 | AAH90998.1 | |||
| BY730334.1 |
More Information
Set Membership: |
|

Genomic Map